Learn More
Sebacoyl chloride, 95%
CAS: 111-19-3 | C10H16Cl2O2 | 239.136 g/mol
Supplier: Thermo Scientific Chemicals A1422622
Description
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.
Solubility
Miscible with hydrocarbons and ethers.
Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.
Specifications
| Sebacoyl chloride | |
| -5°C | |
| 161°C (14 mmHg) | |
| Acrid | |
| 1.468 | |
| 100 g | |
| 1365665 | |
| sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride | |
| WMPOZLHMGVKUEJ-UHFFFAOYSA-N | |
| decanedioyl dichloride | |
| 66072 | |
| 97% |
| 111-19-3 | |
| 1.121 | |
| 165°C (329°F) | |
| C10H16Cl2O2 | |
| MFCD00000770 | |
| UN2922 | |
| Moisture sensitive | |
| Miscible with hydrocarbons and ethers. | |
| C(CCCCC(=O)Cl)CCCC(=O)Cl | |
| 239.136 | |
| 239.14 |
Chemical Identifiers
| 111-19-3 | |
| 239.136 | |
| WMPOZLHMGVKUEJ-UHFFFAOYSA-N | |
| 66072 | |
| C(CCCCC(=O)Cl)CCCC(=O)Cl |
| C10H16Cl2O2 | |
| MFCD00000770 | |
| sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride | |
| decanedioyl dichloride |
Safety and Handling
GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H310-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 203-843-4
RTECSNumber : HD8454250
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only