Learn More
Gibco™ Recharge™ Supplement, Without Glucose and L-glutamine
A chemically defined, animal origin-free (AOF) protein-free supplement formulated without glucose and L-glutamine.
Supplier: Gibco™ 670002
Description
- A chemically defined (CD) supplement designed to increase protein yields, while maintaining acceptable viability and cell growth
- The supplement has been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors
- In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations
- Therefore, these two components should be supplemented by the user and monitored as appropriate
- The Recharge™ Supplement was developed for use in CHO mammalian cell culture systems and can be used either as a media supplement or a culture system feed
- However, user data have shown this CD supplement has been beneficial for non-CHO mammalian systems and microbial systems
- Additionally, the Recharge™ Supplement has been successfully used as a replacement for yeast extract
Spécifications
| Store in cool, dry conditions in well-sealed containers, 2°C to 8°C | |
| CHO Media Supplement | |
| 100 g |
| Powder | |
| Mammalian Cell Culture | |
| Animal origin-free |