missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Recharge™ Supplement, Without Glucose and L-glutamine
GREENER_CHOICE

Catalog No. p-8843013
Change view
Click to view available options
Quantity:
100 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
670004 5 kg
01804551 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 670004 Supplier Gibco™ Supplier No. 670004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A chemically defined, animal origin-free (AOF) protein-free supplement formulated without glucose and L-glutamine.

  • A chemically defined (CD) supplement designed to increase protein yields, while maintaining acceptable viability and cell growth
  • The supplement has been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors
  • In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations
  • Therefore, these two components should be supplemented by the user and monitored as appropriate
  • The Recharge™ Supplement was developed for use in CHO mammalian cell culture systems and can be used either as a media supplement or a culture system feed
  • However, user data have shown this CD supplement has been beneficial for non-CHO mammalian systems and microbial systems
  • Additionally, the Recharge™ Supplement has been successfully used as a replacement for yeast extract

Specifications

Content And Storage Store in cool, dry conditions in well-sealed containers, 2°C to 8°C
Form Powder
Product Type CHO Media Supplement
For Use With (Application) Mammalian Cell Culture
Quantity 5 kg
Source Animal origin-free