Learn More
Corning™ PYREX™ Graham Condensers with Standard Taper Inner and Outer Joints
For use in vacuum distillation
Supplier:  Corning™ 2560500
 Afficher plus de versions de ce produit
Numéro de catalogue. 07-728C
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
For use in vacuum distillation
This condenser has a high surface area per unit length of jacket. Primarily used in vacuum ­distillations. With the outer joint at the top and the inner standard taper joint at the bottom with a drip tip. The tubulation O.D. is approximately 10 mm.
Specifications
Specifications
19.68 in.
For vacuum distillations
Graham Condenser
41 mm
500 mm
1.61 in.
24/40
650 mm
0.39 in.
25.59 in.
Borosilicate Glass
10 mm
Product Suggestions
Product Suggestions