missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PSC Neural Induction Medium

Catalog No. A1647801
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1647801 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1647801 Supplier Gibco™ Supplier No. A1647801
  • ONLINE EXCLUSIVE PRICE
    $1,090.00
    $772.01 / Each

    Save $317.99

    29.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PSC Neural Induction Medium

Gibco™ PSC Neural Induction Medium is a serum-free medium that provides high efficiency neural induction of human pluripotent stems cells (PSCs) in only 7 days. Unlike existing methodologies, use of PSC Neural Induction Medium does not require the intermediary step of embryoid body (EB) formation, which adds time, labor, and variability. Neural Stem Cells (NSCs) generated using PSC Neural Induction Medium have high expression of NSC markers and can be further differentiated into other neural cell types.

PSC Neural Induction Medium enables:

  • Rapid induction of NSCs—high efficiency neural induction in only 7 days, without EB formation
  • Scalable production of NSCs:—generate >20 million NSCs starting from 1 million PSCs
  • Generation of high quality NSCs—resulting NSCs can be cryopreserved, expanded, and differentiated

    Rapid Induction of NSCs
    PSC Neural Induction Medium simplifies the NSC generation workflow by eliminating the need to form EBs or rosettes. By removing this step, PSC Neural Induction Medium enables a more reproducible process with high efficiency neural induction in only 7 days.

    Scalable Production of NSCs
    PSC Neural Induction Medium is a robust medium that has been shown to generate over 20 million NSCs from 1 million PSCs. The medium has been tested with multiple hPSC lines including hiPSCs derived from use of Episomal iPSC Reprogramming Vectors and the CytoTune™-iPS Sendai Reprogramming Kit.

    Generation of High Quality NSCs
    Neural stem cells generated using PSC Neural Induction Medium have >80% expression of Sox1, Sox2, and Nestin (NSC markers), and <1% expression of Oct4 (pluripotency marker). In addition, expanded NSCs maintained phenotype, exhibited normal karyotype, and could be differentiated into neurons, astrocytes, and oligodendrocytes.
  • Specifications

    Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs), Human Stem Cells
    Product Type PSC Neural Induction Media
    Serum Level Serum-free
    Quantity 500 mL
    Content And Storage • 500 mL Basal medium (store at 2–8°C and protect from light)
    • 10 mL Supplement (store at -5 to -20°C and protect from light)
    Can PSC Neural Induction Medium be used to differentiate hPSCs into neurospheres or neurons?

    PSC Neural Induction Medium is a medium to convert hPSCs into NSCs. At day 7 of neural induction, dissociate P0 NSCs with Accutase. The following recommendations can be use for differentiation of neurons or neurospheres: For differentiation of neurons: Use PSC Neural Induction Medium to convert hPSCs into NSCs. At day 7 of neural induction, dissociate P0 NSCs with Accutase. If dissociated NSCs are plated on a Geltrex-coated culture vessel in neural expansion medium, NSCs will grow as a mono-layer. For NSC expansion, it is necessary to treat NSCs with 5 µM ROCK inhibitor Y27632 at the time of plating to prevent cell death if NSCs are under P4. For differentiation of neurospheres: Use PSC Neural Induction Medium to convert hPSCs into NSCs. At day 7 of neural induction, dissociate P0 NSCs with Accutase. If dissociated NSCs are plated into a non-coated flask in neural expansion medium, dissociated NSCs will re-aggregate into small spheres. NSCs in each sphere will proliferate and form big neurospheres. We have not expanded NSCs in neurosphere format in-house. You can test expanding NSCs in neurospheres with and without ROCK inhibitor Y27632 to determine which is optimal for your workflow. Neural stem cells in adherent or neurosphere conditions can be differentiated into neurons using the appropriate protocol.

    Have you performed flow cytometry or qPCR studies to determine the expression of NKX2.1 in NSCs generated from iPSC using PSC Neural Induction Medium?

    Studies have not been performed using flow cytometry or qPCR to detect NKX2.1 expression in NSCs induced by Neural Induction Medium.

    Is it normal to see loss of pluripotency markers (i.e. SSEA-4, Oct-4, and Tra1-60) and still test positive for SOX2 expression when NSCs are cultured in neural expansion medium?

    Yes, this is normal. SOX2 is expressed by both hPSCs and hPSC-derived NSCs, so you will see its expression in the induced NSCs from iPSCs.

    Can NSCs induced by PSC Neural Induction Medium be differentiated into neurons on glass slides/coverslips coated with laminin and poly-L-ornithine?

    NSCs induced using the PSC Neural Induction Medium can be differentiated on glass slides/coverslips, but the glass should be cleaned well before coating. We recommend the following cleaning protocol:
    1. Treat glass cover slips with1M HCl at RT for 2-3 h on a shaker.
    2. Rinse with tap water 5 times, and then double distilled water for 2 times
    3. Store cleaned cover slips in 70% ethanol.
    4. Dry cover slips by transferring cover slips into culture plate with sterile forceps before poly-L-ornithine and laminin coating.

    Can I maintain differentiated neurospheres in Neurobasal+B27+N2+bFGF+EGF?

    Neurospheres can be plated on laminin coated culture plates for neuron differentiation. The issue is that it is difficult to control the plating density of neurospheres. Alternatively, neurospheres can be dissociated into single cells and plate single cell suspension at a certain density such as 1-5 x 10^4 cells/cm2 onto laminin coated plates for neuron differentiation. For general neuron differentiation, Neurobasal+B27+N2 can be used. Growth factors such as BDNF and/or GDNF can be added into medium for improving survival of differentiating NSCs.

    Can I maintain neurospheres in PSC Neural Expansion Medium?

    Yes, neurospheres can be maintained in PSC Neural Expansion medium. The recipe for Neural Expansion medium is found in the Neural Induction Medium product manual.

    Why am I seeing abnormal staining pattern of NSCs stained with antibodies against neural markers?

    Antibodies that are not stored or handled properly may result in loss of staining quality and should be replaced. Note that Nestin should stain filaments in the cytoplasm whereas Pax6, Sox1, and Sox2 should stain the nucleus. Oct4 should stain the nucleus of pluripotent cells, but not NSCs.

    What should I do if I see extensive cell death after thawing and plating cryo-preserved NSCs?

    If you are seeing extensive cell death and the NSCs are cryo-preserved under P4, we recommend treating with 5 µM ROCK Inhibitor Y27632 after plating.

    Can NSCs induced by PSC Neural Induction Medium be differentiated into motor neurons?

    Yes, NSCs induced by PSC Neural Induction Medium can be differentiated into motor neurons. We do not offer a validated protocol, but you can refer to the following publication or perform a literature search for additional methods:
    http://rd.springer.com/article/10.1007/s12015-014-9541-0

    What PSC cell lines have been tested using the PSC Neural Induction Medium kit?

    We have tested the following PSC cell lines for the PSC Neural Induction Medium:
    • H9 embryonic stem cells
    • iPSC line created in-house
    • Episomal iPSC cell line

    Can NSCs generated using PSC Neural Induction Medium be expanded in the PSC Neural Induction Medium or do they need to be in the recommended NSC expansion medium, as stated in the product manual?

    We recommend using neural expansion medium with the following composition to expand NSCs generated using the PSC Neural Induction Medium: - 50:50 Neurobasal medium: Advanced DMEM/F12 plus PSC Neural Induction Supplement.

    PSC lines can behave differently, and we recommend performing your own validation experiments if you want to use PSC Neural Induction Medium, or you do not observe good expansion using the Neurobasal and Advanced DMEM/F12 combination.

    When should I add ROCK inhibitor to PSC Neural Induction Medium?

    It is not necessary or recommended to add ROCK inhibitor to PSC Neural Induction Medium when differentiating cells into neural stem cells. You would only add ROCK inhibitor to iPSC or ESC before initiation of neural induction and when expanding neural stem cells using Neural Expansion Medium.

    Can NSCs derived by PSC Neural Induction Medium be differentiated into cells in the peripheral nervous system (PNS)?

    No, NSCs derived by PSC Neural Induction Medium cannot be differentiated into cells of the PNS. They can be differentiated into neurons and glial cells in the central nervous system. Sensory, sympathetic and parasympathetic neurons, as well as Schwann cells in the PNS are derived from precursor cells of the neural crest.

    Can PSC Neural Induction Medium convert mouse PSCs into NSCs?

    PSC Neural Induction Medium has not been tested and is not recommended for the conversion of mouse PSCs into NSCs.

    What should I do if my NSCs reach near maximal confluence within 2-3 days after passaging?

    NSCs derived from some human PSC lines may have an increased proliferation rate. In this case, we recommend decreasing NSC plating density to 5x10^4 cells/cm2.

    What should I do if I see extensive cell death after plating dissociated NSCs for expansion and differentiation?

    We recommend adding ROCK inhibitor Y27632 into cell suspension to reach a final concentration of 5 µM if the passage number of NSCs is under P4. NSCs from some human PSC lines may be more sensitive to dissociation. For NSCs derived from those PSC lines, the overnight treatment with Y27632 after P4 decreases cell death after re-plating.

    How can I prevent extensive cell death during the late stage of neural induction (Day 5-7) with PSC Neural Induction Medium?

    To prevent extensive cell death during late stage neural induction, we recommend checking whether cells are over-confluent. If they are, change the PSC Neural Induction Medium every day with 5 mL per well of a 6-well plate.

    How can I prevent cell detachment during neural induction with PSC Neural Induction Medium?

    For PSCs cultured in Essential 8 Medium, cells may detach during neural induction. To prevent cell detachment, we recommend coating your culture plates with Vitronectin at 1µg/cm2 when splitting PSCs.

    How can I remove non-neural cells during NSC expansion following the induction of NSCs from hPSCs using PSC Neural Induction Medium?

    We recommend that you discard the culture and begin the process again. You will want to select and maintain a high quality of PSCs before inducing neural differentiation.

    How do I get the right starting density for my human PSCs?

    Due to variable parameters such as the confluence of PSCs, cell clump passaging, cell attachment and the property of different PSC lines, it may be difficult to determine the splitting ratio. To obtain the optimal starting density for neural induction with PSC Neural Induction Medium we recommend estimating the cell number of PSC clumps before plating. Instructions for estimating the cell number of PSC clumps can be found in the protocol (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0008031.pdf). PSC culture should be at 15-25% confluence on day 1 of PSC splitting (Day 0 of neural induction).

    Which format of cultures should I use for neural induction with PSC Neural Induction Medium?

    Neural induction can be started with human PSCs cultured on 6-well plate or culture dishes. PSCs cultured in flasks are not recommended because it is difficult to remove non-neural differentiated colonies in flasks.

    Does PSC Neural Induction Medium work for human PSCs cultured on feeders (mouse embryonic fibroblasts, MEF) and feeder-free conditions?

    PSC Neural Induction Medium has been tested on human PSCs cultured in both feeder-free and feeder-based conditions. To eliminate MEF contamination, human PSCs cultured in feeder-free conditions such as Essential 8 and StemPro hESC SFM are strongly recommended for starting neural induction.

    Does PSC Neural Induction Medium work for both human ESCs and iPSCs?

    Yes, PSC Neural Induction Medium works for both human ESCs and iPSCs.

    Does PSC Neural Induction Medium (Cat. No. A1647801) contain any SMAD inhibitors, retinoic acid, or anything that directs differentiation?

    Unfortunately, we cannot provide the exact composition of PSC Neural Induction Medium because it is proprietary information. The only information we can provide is that the medium contains BSA, small molecules, and products capable of inhibiting GSK and TGF-beta pathways.

    Is StemFlex Medium compatible with downstream differentiation kits available from Thermo Fisher Scientific?

    Yes. We have seen compatibility with the following differentiation kits provided by Thermo Fisher Scientific: PSC Cardiomyocyte Differentiation Kit (Cat. No. A2921201), PSC Definitive Endoderm Induction Kit (Cat. No. A3062601), PSC Neural Induction Medium (Cat. No. A1647801), and PSC Dopaminergic Neuron Differentiation Kit (Cat. No. A3147701).

    Why did my Gibco PSC Neural Induction Medium (Cat. No. A1647801) fail to induce neural stem cells?

    There are several possibilities causing the failure of neural induction:

    - High quality of hPSC cells is critical to the success of neural induction. Remove differentiated and partially differentiated hPSC cells before neural induction.
    - Before plating hPSCs for induction, cell counting is recommended because too low or too high cell confluency will reduce induction efficiency. The recommended plating density for induction is 2-2.5 x 10E4 cells/cm2.
    - Cell clumps (and not a single cell suspension) should be plated for induction.
    - To increase induction efficiency, overnight treatment with 10 µM ROCK Inhibitor Y27632 at the time hPSC splitting can be used to prevent extensive cell death.

    After neural induction from iPSC to NSCs, can I use Gibco StemPro NSC SFM (Cat. No. A1050901) to expand NSCs?

    We do not recommend using Gibco StemPro NSC SFM to expand NSCc which are induced using Gibco PSC Neural Induction Medium (Cat. No. A1647801), because the morphology of NSCs induced from some lines of hPSCs will change if this medium is used to expand NSCs. We recommend using the Neural Expansion Medium to expand such NSCs, as stated here ( https://tools.thermofisher.com/content/sfs/manuals/MAN0008031.pdf).

    Can I use PSCs that are long-term cultured in Gibco Essential 8 Flex Medium for downstream differentiation?

    We have seen no effect on downstream differentiation potential for cells cultured in Essential 8 Flex Medium for up to 15 passages. Tri-lineage potential has been demonstrated from embryoid bodies as well as using Gibco PSC Neural Induction Medium (Cat. No. A1647801), Gibco PSC Cardiomyocyte Differentiation Kit (Cat. No. A25042SA) and Gibco PSC Definitive Endoderm Induction Kit (Cat. No. A27654SA).


    For Research Use Only. Not for use in diagnostic procedures.