Learn More
Promega Restriction Enzymes - Pvu I (D)
Active and capable of digestion directly in GoTaq™ Green Master Mix. (CG AT/CG GC\TA GC)
Supplier: Promega R6325
Description
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
Greener Choice Claims
- Environmental benefits include:
- Énergie renouvelable
Spécifications
2 to 10 U/μL | |
37°C | |
Active and capable of digestion directly in GoTaq™ Green Master Mix | |
-30°C to -10°C | |
CGAT.CG | |
Liquid |
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U PvuI | |
7.3 | |
10mM Tris HCl (pH 7.3 at 25°C), 50mM NaCl, 0.1% Triton™ X-100, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol | |
500 U | |
PvuI |