Learn More
Promega ONE-Glo™ Luciferase Assay System DFS Item
The ONE-Glo™ Luciferase Assay System provides a sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells. Ideal for high- and ultrahigh-throughput applications.
Supplier:  Promega E6130
 Afficher plus de versions de ce produit
Numéro de catalogue. PRE6130
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Découvrez plus d'offres spéciales
Comprend: ONE-Glo Luciferase Assay Buffer, ONE-Glo Luciferase Assay Substrate
Description
Description
  • Robust Performance for High- and Ultrahigh-Throughput Reporter Assays
  • Less sensitive to mixing and dispensing conditions, enhancing reproducibility
  • Ideal for use in high-density (384- and 1536-well) microplates
  • Reduced odor, and convenient storage options
Spécifications
Spécifications
1 x 1000mL ONE-Glo™ Luciferase Assay Buffer, 1 x 1 vial ONE-Glo™ Luciferase assay substrate
6.7
-30°C to -10°C
1000 mL
Luminescence
384- and 1536-well plates
Detection of firefly luciferase reporter gene expression in mammalian cells
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
  • Patent pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  • Certain applications of this product may require licenses from others.