missing translation for 'onlineSavingsMsg'
Learn More

Promega ONE-Glo™ Luciferase Assay System

Catalog No. p-3606288
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE6110 10 mL
PRE6130 1000 mL
PRE6120 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRE6110 Supplier Promega Supplier No. E6110
Only null left

Includes

ONE-Glo Luciferase Assay Buffer, ONE-Glo Luciferase Assay Substrate

The ONE-Glo™ Luciferase Assay System provides a sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells. Ideal for high- and ultrahigh-throughput applications.

  • Robust Performance for High- and Ultrahigh-Throughput Reporter Assays
  • Less sensitive to mixing and dispensing conditions, enhancing reproducibility
  • Ideal for use in high-density (384- and 1536-well) microplates
  • Reduced odor, and convenient storage options

Specifications

Includes 1 x 10mL ONE-Glo™ Luciferase Assay Buffer, 1 x 1 vial ONE-Glo™ Luciferase assay substrate
Detection Method Luminescence
pH 6.7
For Use With (Equipment) 384- and 1536-well plates
Content And Storage -30°C to -10°C
For Use With (Application) Detection of firefly luciferase reporter gene expression in mammalian cells
Quantity 10 mL

  • Patent pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  • Certain applications of this product may require licenses from others.