missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKACG Mouse anti-Human, Clone:2E4, Abnova™
Mouse monoclonal antibody raised against recombinant human PRKACG.
Supplier: Abnova Corporation MAB17724
Description
Cyclic AMP-dependent protein kinase (PKA) consists of two catalytic subunits and a regulatory subunit dimer. This gene encodes the gamma form of its catalytic subunit. The gene is intronless and is thought to be a retrotransposon derived from the gene for the alpha form of the PKA catalytic subunit. [provided by RefSeq]Spécifications
PRKACG | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
PRKACG | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin) | |
2E4 | |
protein kinase, cAMP-dependent, catalytic, gamma | |
In ascites (0.03% sodium azide) | |
KAPG, PKACg | |
Recombinant protein corresponding to human PRKACG from E. coli. | |
5568 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |