Learn More
MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Replacement Glass Columns
Replacement glass columns are available in a wide range of bore sizes and lengths.
Supplier:  MilliporeSigma™ Supelco™ K5171U
 Afficher plus de versions de ce produit
Numéro de catalogue. 111009271
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Replacement columns are available to replace broken columns. The modular nature of Benchmark columns provides ultimate flexibility. Different column sizes can be mixed and matched with different endpiece types to meet specific requirements.
Spécifications
Spécifications
35 mm
250 mm
Omnifit
Chromatography
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Omnifit is a registered trademark of Diba Industries