Learn More
MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Replacement Glass Columns
Replacement glass columns are available in a wide range of bore sizes and lengths.
$310.61 - $1051.42
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
|
111010280
|
MilliporeSigma™ Supelco™
K5162U |
250 mm | 15 mm |
Each for $310.61
|
|
||||
|
111010192
|
MilliporeSigma™ Supelco™
K5163U |
330 mm | 15 mm |
Each for $312.54
|
|
||||
|
111010132
|
MilliporeSigma™ Supelco™
K5164U |
400 mm | 15 mm |
Each for $326.18
|
|
||||
|
111009745
|
MilliporeSigma™ Supelco™
K5168U |
330 mm | 25 mm |
Each for $437.12
|
|
||||
|
111009721
|
MilliporeSigma™ Supelco™
K5169U |
400 mm | 25 mm |
Each for $440.99
|
|
||||
|
111009366
|
MilliporeSigma™ Supelco™
K5170U |
150 mm | 35 mm |
Each for $574.14
|
|
||||
|
111009271
|
MilliporeSigma™ Supelco™
K5171U |
250 mm | 35 mm |
Each for $455.37
|
|
||||
|
111009198
|
MilliporeSigma™ Supelco™
K5172U |
330 mm | 35 mm |
Each for $641.04
|
|
||||
|
111009153
|
MilliporeSigma™ Supelco™
K5173U |
400 mm | 35 mm |
Each for $691.09
|
|
||||
|
111008607
|
MilliporeSigma™ Supelco™
K5176U |
160 mm | 50 mm |
Each for $1,051.42
|
|
||||
|
111008681
|
MilliporeSigma™ Supelco™
K5174U |
250 mm | 50 mm |
Each for $978.20
|
|
||||
|
111008626
|
MilliporeSigma™ Supelco™
K5175U |
330 mm | 50 mm |
Each for $1,030.37
|
|
||||
|
111009981
|
MilliporeSigma™ Supelco™
K5153U |
330 mm | 6.6 mm |
Each for $376.39
|
|
||||
|
111009910
|
MilliporeSigma™ Supelco™
K5154U |
400 mm | 6.6 mm |
Each for $401.99
|
|
||||
Description
Replacement columns are available to replace broken columns. The modular nature of Benchmark columns provides ultimate flexibility. Different column sizes can be mixed and matched with different endpiece types to meet specific requirements.Specifications
| Omnifit | |
| Chromatography |
Omnifit is a registered trademark of Diba Industries