missing translation for 'onlineSavingsMsg'
Learn More

Mesityl oxide, 99%, mixture of alpha- and beta-isomers

CAS: 141-79-7 | C6H10O | 98.14 g/mol

Supplier:  Thermo Scientific Chemicals 125600100

 Afficher plus de versions de ce produit

Numéro de catalogue. AC125600100


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications
Afficher plus de résultats
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 205-502-5

Recommended Storage : Product may darken during storage

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only