missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylyl Cyanide 98.0+%, TCI America™
Supplier: TCI America X004625ML
Spécifications
m-Xylyl Cyanide | |
Yellow | |
C9H9N | |
25 mL | |
2-3-methylphenyl acetonitrile, 3-methylbenzyl cyanide, m-tolylacetonitrile, m-methylphenylacetonitrile, m-xylyl cyanide, 3-methylbenzylcyanide, m-methylbenzyl cyanide, 3-methylphenylacetonitrile, 3-methylphenyl acetonitrile, 2-m-tolyl acetonitrile | |
CC1=CC=CC(CC#N)=C1 | |
131.18 | |
131.18 | |
Liquid |
2947-60-6 | |
130°C | |
MFCD00001914 | |
3276 | |
WOJADIOTNFDWNQ-UHFFFAOYSA-N | |
2-(3-methylphenyl)acetonitrile | |
76279 | |
≥98.0% (GC) |
Identifiants chimiques
2947-60-6 | |
131.18 | |
WOJADIOTNFDWNQ-UHFFFAOYSA-N | |
76279 | |
CC1=CC=CC(CC#N)=C1 |
C9H9N | |
MFCD00001914 | |
2-3-methylphenyl acetonitrile, 3-methylbenzyl cyanide, m-tolylacetonitrile, m-methylphenylacetonitrile, m-xylyl cyanide, 3-methylbenzylcyanide, m-methylbenzyl cyanide, 3-methylphenylacetonitrile, 3-methylphenyl acetonitrile, 2-m-tolyl acetonitrileAfficher plus de résultats | |
2-(3-methylphenyl)acetonitrile |
Sécurité et manipulation
missing translation for 'tsca' : No