missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Erythrulose

CAS: 533-50-6 | C4H8O4, C4H8O4 | 120.10 g/mol

Supplier:  Thermo Scientific Chemicals 467301000

 View more versions of this product

Catalog No. AC467301000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Erythrulose
100 g
110°C
120.12
UQPHVQVXLPRNCX-YEQUQEJONA-N
(3S)-1,3,4-trihydroxybutan-2-one
533-50-6
Solubility aq. soln.: soluble.
C4H8O4
MFCD00004703
OC[C@H](O)C(=O)CO
120.10

Chemical Identifiers

533-50-6
C4H8O4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only