missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Erythrulose

Catalog No. p-7214693
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467300250 25 g
AC467301000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467300250 Supplier Thermo Scientific Chemicals Supplier No. 467300250
Only null left

CAS: 533-50-6 | C4H8O4, C4H8O4 | 120.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 533-50-6
Molecular Formula C4H8O4

Specifications

Quantity 25 g
Solubility Solubility aq. soln.: soluble.
Flash Point 110°C
Formula Weight 120.12
MDL Number MFCD00004703
InChI Key UQPHVQVXLPRNCX-YEQUQEJONA-N
SMILES OC[C@H](O)C(=O)CO
IUPAC Name (3S)-1,3,4-trihydroxybutan-2-one
Molecular Weight (g/mol) 120.10
Chemical Name or Material L-Erythrulose

RUO – Research Use Only