Learn More
DWK Life Sciences Kimble™ Adapters for Round-Bottom Centrifuge Tubes
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
Supplier: DWK Life Sciences 4555015
Description
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
- Made of rubber
Greener Choice Claims
- Environmental benefits include:
- Énergie renouvelable
- Recyclable
- Non toxique
Spécifications
Centrifuge Tube Adapter | |
Rubber | |
15 mL Round-bottom Centrifuge Tube |