Learn More
DWK Life Sciences Kimble™ Adapters for Round-Bottom Centrifuge Tubes
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
$198.82 - $229.82
| Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
|
0556911
|
DWK Life Sciences
4555030 |
30 mL Round-bottom Centrifuge Tube |
Case of 2 Each for $229.82
|
|
|||||
|
0556910
|
DWK Life Sciences
4555015 |
15 mL Round-bottom Centrifuge Tube |
Case of 2 Each for $198.82
|
|
|||||
Description
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
- Made of rubber
Specifications
| Rubber |