missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose-Glutamic Acid Solution, Standard Check Solution for Biochemical Oxygen Demand (BOD), Ricca Chemical
Supplier: Ricca Chemical Company 325532
Spécifications
Glucose-Glutamic Acid Solution | |
Amber Glass Bottle | |
98.0,0.02,0.01 | |
Liquid | |
Colorless | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
1 L | |
CHEBI:42758 |
Standard Check Solution for Biochemical Oxygen Demand (BOD) | |
50-99-7,56-86-0,7732-18-5 | |
100.0,0.02,0.02 | |
C6H12O6 | |
100°C | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.156 | |
107526 |
Identifiants chimiques
50-99-7 | |
180.156 | |
107526 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
C6H12O6 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
CHEBI:42758 | |
C(C(C(C(C(C=O)O)O)O)O)O |