missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Emtricitabine

CAS: 143491-57-0 | C8H10FN3O3S

Supplier:  Thermo Scientific Chemicals 462070050

 Afficher plus de versions de ce produit

Numéro de catalogue. AC462070050


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Emtricitabine
5 g,5 g
153622
C8H10FN3O3S
143491-57-0
Solubility in water: soluble. Other solubilities: very soluble in methanol,very slightly soluble in acetonitrile
136°C to 140°C
247.24

Identifiants chimiques

143491-57-0
C8H10FN3O3S
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only