missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Emtricitabine

CAS: 143491-57-0 | C8H10FN3O3S

$627.65 - $627.65

Chemical Identifiers

CAS 143491-57-0
Molecular Formula C8H10FN3O3S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC462070050
View Documents
Thermo Scientific Chemicals
462070050
5 g
Each for $627.65
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

143491-57-0
C8H10FN3O3S

Specifications

143491-57-0
Solubility in water: soluble. Other solubilities: very soluble in methanol,very slightly soluble in acetonitrile
136°C to 140°C
247.24
5 g,5 g
153622
C8H10FN3O3S
Emtricitabine
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only