Learn More
MilliporeSigma™ Supelco™ Empty Tubes for Thermal Desorption
Supplier: MilliporeSigma™ Supelco™ 28714U
Spécifications
| (10) Tension Springs | |
| Non-Fritted | |
| Thermal Desorption Tube | |
| Glass | |
| 3.5 in. |
| 10/Pk. | |
| Packing Adsorbents, Sample Preparation Applications | |
| 1/4 in. | |
| PerkinElmer, Markes, DANI, OI Analytical, Shimadzu Instruments |