Learn More
MilliporeSigma™ SeQuant™ ZIC™-pHILIC Polymeric Analytical PEEK Columns
PEEK coated HPLC column
Supplier:  MilliporeSigma™ 1504630001
 Afficher plus de versions de ce produit
Numéro de catalogue. M1504630001
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
The polymeric SeQuant™ ZIC™-pHILIC columns is designed for particularly difficult separations of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase is attached to porous polymer beads and tolerates pH from 2-11.
Spécifications
Spécifications
4.6 mm
50 mm
Column
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS