Learn More
DuPont™ Tyvek™ 400 Lab Coats, Open Wrists
GREENER_CHOICE
Limited-use lab coats offer easy donning and doffing and provide an ideal balance of protection, durability, and comfort.
Supplier:  DuPont™ TY212SWHSM003000
 Afficher plus de versions de ce produit
Numéro de catalogue. 01-361-10A
Il en reste null
Découvrez plus d'offres spéciales
Avertissement:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
Limited-use lab coats offer easy donning and doffing and provide an ideal balance of protection, durability, and comfort.
  • Provides protection against submicron particles
  • Lightweight and tear-resistant Tyvek fabric
  • Knee length with five-snap front and full sleeves
  • Limited splash protection
  • Ideal for lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up
  • Easy donning and doffing
  • Laydown collar, open wrists
  • Two front pockets for holding writing instruments, small tools or parts
Compliance
Meets ANSI/ISEA 101-1996 standards.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Spécifications
Spécifications
General Purpose Lab Coat
Unisex
White
Open
Non-sterile
2
TY212SWH
32.25 to 35.75 in.
Bulk Packaged
77.47 cm
Small
Tyvek
Snaps
Limited-use
Below Hip-length
Lay Down
Serged
Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning
30.5 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS