Learn More
Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile
Specially designed Netwell™ carriers with handles allow simultaneous processing of up to 12 samples per carrier
Supplier:  Corning™ C3517
 Afficher plus de versions de ce produit
Numéro de catalogue. 07-200-221
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Specially designed Netwell carriers and handles allow simultaneous processing of up to 12 samples per carrier
  • Polystyrene reagent trays are available in white for colorimetric reaction contrast, or black for better visibility of tissue sections
  • Each carrier kit contains eight carriers and eight handles
Spécifications
Spécifications
Tray
25/Pk.
200/Cs.
Black
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS