Learn More
Corning™ Filtered IsoTip™ Universal Fit Racked Pipet Tips
GREENER_CHOICE
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
Supplier:  Corning™ 4823
 Afficher plus de versions de ce produit
Numéro de catalogue. 07-200-267
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
  • Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
  • Packaged sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Greener Choice Claims
  • Environmental benefits include:
    • Énergie renouvelable
Spécifications
Spécifications
1 to 200 μL
Sterile
Rack
Rack
Yes
Polypropylene
Filtered
Natural
Round
Universal Fit
Isotip
10 μL, 50 μL, 100 μL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS