Learn More
Cesium nitrate, 99.8% (metals basis)
Preparation of Cs compounds | CAS: 7789-18-6 | CsNO3 | 194.909 g/mol
Supplier: Thermo Scientific Chemicals 01288430
Description
Preparation of cesium compoundsCesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| (decomposition) | |
| 414°C | |
| Crystalline | |
| 99.8% (metals basis) | |
| MFCD00010963 | |
| Soluble in 5 parts cold and 0.5 parts boiling water. Soluble in acetone. Slightly soluble in alcoholSoluble in water and acetone. Slightly soluble in ethanol. | |
| [N+](=O)([O-])[O-].[Cs+] | |
| 194.909 | |
| 194.91 | |
| Hygroscopic |
| Cesium nitrate | |
| 7789-18-6 | |
| 250 g | |
| CsNO3 | |
| cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
| NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
| cesium;nitrate | |
| 62674 | |
| 1.55 | |
| 3.68 |
Identifiants chimiques
| 7789-18-6 | |
| 194.909 | |
| NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
| 62674 | |
| [N+](=O)([O-])[O-].[Cs+] |
| CsNO3 | |
| MFCD00010963 | |
| cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
| cesium;nitrate |
Sécurité et manipulation
P201-P202-P220-P221-P264-P270-P272-P280-P301+P312-P308+P313-P330-P370+P378q-P370+P380+P375-P372-P501c
H271-H302-H361f
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: III
missing translation for 'einecsNumber' : 232-146-8
missing translation for 'rtecsNumber' : FL0700000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only

