missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 99%, for biochemistry

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

Supplier:  Thermo Scientific Chemicals 172622500

 Afficher plus de versions de ce produit

Numéro de catalogue. AC172622500


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Spécifications

Spécifications

0.04 max. at 260nm
99%
1135-40-6
Powder
99%
5% max.
C6H11NH(CH2)3SO3H
Glass bottle
3-Cyclohexylamino-1-propanesulfonic acid
OS(=O)(=O)CCCNC1CCCCC1
221.32
221.31
Biochemical
CAPS
(0.1M in H2O)
White
250 g
Authentic
C9H19NO3S
MFCD00003837
15, 1769
PJWWRFATQTVXHA-UHFFFAOYSA-N
3-(cyclohexylamino)propane-1-sulfonic acid
70815
≥98.5% (on dry substance)

Identifiants chimiques

1135-40-6
221.32
PJWWRFATQTVXHA-UHFFFAOYSA-N
70815
OS(=O)(=O)CCCNC1CCCCC1
C9H19NO3S
MFCD00003837
3-Cyclohexylamino-1-propanesulfonic acid
3-(cyclohexylamino)propane-1-sulfonic acid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 214-492-1

missing translation for 'rtecsNumber' : TZ6395000

missing translation for 'tsca' : TSCA

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only