missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 99%, for biochemistry

Catalog No. p-81983
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172622500 250 g Glass bottle
AC172620010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172622500 Supplier Thermo Scientific Chemicals Supplier No. 172622500
Only null left

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym 3-Cyclohexylamino-1-propanesulfonic acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Absorbance 0.04 max. at 260nm
Absorbance Conditions (0.1M in H2O)
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 5% max.
Linear Formula C6H11NH(CH2)3SO3H
Packaging Glass bottle
Merck Index 15, 1769
Formula Weight 221.31
Percent Purity ≥98.5% (on dry substance)
Grade Biochemical
Chemical Name or Material CAPS, 99%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-492-1
RTECSNumber TZ6395000
TSCA TSCA

RUO – Research Use Only