Learn More
Bel-Art™ Slide Boxes
Will not warp, splinter or crack, and are unaffected by moisture. Slide boxes are compact, durable slide boxes to provide the maximum protection for 25 x 75 mm (1 x 3 in.) microscope slides.
Supplier:  Bel-Art™ H445760000
 Afficher plus de versions de ce produit
Numéro de catalogue. 14-512-37
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Heavy-walled boxes are rugged and will not bend, split or chip
  • Boxes are unaffected by humidity and moisture and are thoroughly insect-proof
  • Easily identify slides with numbered index included on base and cover
Spécifications
Spécifications
Slide Box
Detachable
Microscope Slide
25
25
Numbered index on base
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS