Learn More
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, 97%
CAS: 77-08-7 | C21H24O4 | 340.419 g/mol
Supplier: Thermo Scientific Chemicals B2217018
Description
5,5',6,6'-Tetrahydroxy-3,3,3',3'- tetramethyl-1,1'-spirobisindane is used in the preparation of 4,7,4',7'-tetrabromo-3,3,3',3'-tetramethyl-2,3,2',3'-tetrahydro-[1,1']spirobiindene-5,6,5',6'-tetraol by bromination using bromine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5,5′,6,6′-Tetrahydroxy-3,3,3′,3′- tetramethyl-1,1′-spirobisindane is used in the preparation of 4,7,4′,7′-tetrabromo-3,3,3′,3′-tetramethyl-2,3,2′,3′-tetrahydro-[1,1′]spirobiindene-5,6,5′,6′-tetraol by bromination using bromine.
Solubility
Slightly soluble in methanol.
Notes
Incompatible with strong oxidizing agents.
Spécifications
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane | |
>300°C | |
MFCD00021235 | |
2226231 | |
Slightly soluble in methanol. | |
CC1(CC2(CC(C3=CC(=C(C=C32)O)O)(C)C)C4=CC(=C(C=C41)O)O)C | |
340.419 | |
340.42 |
77-08-7 | |
C21H24O4 | |
50 g | |
5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, tts 5, 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindan, 1,1'-spirobi 1h-indene-5,5',6,6'-tetrol, 2,2',3,3'-tetrahydro-3,3,3',3'-tetramethyl, 1,1'-spirobi indane-5,6-diol , 3,3,3',3'-tetramethyl, 1,1,1',1'-tetramethyl-3,3'-spirobi 2h-indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2h,2'h-1,1'-spirobi indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2,2',3,3'-tetrahydro-1,1'-spirobi indene-5,5',6,6'-tetrol, 1,6-diol , 3,3,3',3'-tetramethyl | |
POFMQEVZKZVAPQ-UHFFFAOYSA-N | |
1,1,1',1'-tetramethyl-3,3'-spirobi[2H-indene]-5,5',6,6'-tetrol | |
66162 | |
97% |
Identifiants chimiques
77-08-7 | |
340.419 | |
POFMQEVZKZVAPQ-UHFFFAOYSA-N | |
66162 | |
CC1(CC2(CC(C3=CC(=C(C=C32)O)O)(C)C)C4=CC(=C(C=C41)O)O)C |
C21H24O4 | |
MFCD00021235 | |
5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, tts 5, 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindan, 1,1'-spirobi 1h-indene-5,5',6,6'-tetrol, 2,2',3,3'-tetrahydro-3,3,3',3'-tetramethyl, 1,1'-spirobi indane-5,6-diol , 3,3,3',3'-tetramethyl, 1,1,1',1'-tetramethyl-3,3'-spirobi 2h-indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2h,2'h-1,1'-spirobi indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2,2',3,3'-tetrahydro-1,1'-spirobi indene-5,5',6,6'-tetrol, 1,6-diol , 3,3,3',3'-tetramethylAfficher plus de résultats | |
1,1,1',1'-tetramethyl-3,3'-spirobi[2H-indene]-5,5',6,6'-tetrol |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 201-003-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only