missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Supplier: Hamilton™ HAM766101
Description
- PTFE coating from point for 0.75 in.
- For dispensing both liquids and gases
- Precision-machined polymer plunger tip that creates leak-free seal
Available as replacement needle for installing into standard removable needle (RN) syringe; Needles sold separately
Spécifications
| Autoclavable | |
| 26s G | |
| 1805 | |
| 50 μL |