Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
$97.83 - $380.12
Products 18
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Model Price Quantity  
14-824-680
View Documents
Hamilton™
HAM765801
500 μL 1750
Each for $224.59
Only null left
14-824-679
View Documents
Hamilton™
HAM765701
250 μL 1725
Each for $133.42
Only null left
14-824-665
View Documents
Hamilton™
HAM763801
100 μL 710
Each for $170.77
Only null left
14-824-666
View Documents
Hamilton™
HAM763901
250 μL 725
Each for $289.00
Only null left
14-824-667
View Documents
Hamilton™
HAM764001
500 μL 750
Each for $322.85
Only null left
14-824-662
View Documents
Hamilton™
HAM763501
10 μL 701
Each for $97.83
Only null left
14-824-663
View Documents
Hamilton™
HAM763601
25 μL 702
Each for $196.88
Only null left
14-824-664
View Documents
Hamilton™
HAM763701
50 μL 705
Each for $232.91
Only null left
14-824-714
View Documents
Hamilton™
HAM765201
10 mL 1010
Each for $218.79
Only null left
14-824-688
View Documents
Hamilton™
HAM765101
5 mL 1005
Each for $291.62
Only null left
14-824-685
View Documents
Hamilton™
HAM766301
250 μL 1825
Each for $380.12
Only null left
14-824-686
View Documents
Hamilton™
HAM764901
1 mL 1001
Each for $257.88
Only null left
14-824-687
View Documents
Hamilton™
HAM765001
2.5 mL 1002
Each for $172.55
Only null left
14-824-683
View Documents
Hamilton™
HAM766101
50 μL 1805
Each for $367.12
Only null left
14-824-682
View Documents
Hamilton™
HAM766001
25 μL 1802
Each for $235.51
Only null left
14-824-684
View Documents
Hamilton™
HAM766201
100 μL 1810 RN
Each for $369.71
Only null left
14-824-681
View Documents
Hamilton™
HAM765901
10 μL 1801 RN
Each for $356.71
Only null left
14-824-651
View Documents
Hamilton™
763401
5 μL 75RN
Each for $123.60
Only null left
Description
Description
Gastight syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.