missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl alcohol, 98%

CAS: 105-13-5 | C8H10O2 | 138.17 g/mol

$71.16 - $245.86

Identifiants chimiques

CAS 105-13-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004653
InChI Key MSHFRERJPWKJFX-UHFFFAOYSA-N
Synonym 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanolAfficher plus de résultatsAfficher moins de résultats
PubChem CID 7738
ChEBI CHEBI:86918
IUPAC Name (4-methoxyphenyl)methanol
SMILES COC1=CC=C(CO)C=C1
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC159731000
FDS Afficher les documents
Thermo Scientific Chemicals
159731000
100 g Glass bottle
chaque for $71.16
Il en reste null
 
AC159735000
FDS Afficher les documents
Thermo Scientific Chemicals
159735000
500 g Glass bottle
chaque for $245.86
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications
Afficher plus de résultats
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-273-6

missing translation for 'rtecsNumber' : DO8925000

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only