missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanophenol, 99%

CAS: 611-20-1 | C7H5NO | 119.12 g/mol

87,74 $ - 208,34 $

Identifiants chimiques

CAS 611-20-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002145
InChI Key CHZCERSEMVWNHL-UHFFFAOYSA-N
Synonym 2-cyanophenol, o-cyanophenol, salicylonitrile, o-hydroxybenzonitrile, benzonitrile, 2-hydroxy, salicylnitrile, benzonitrile, hydroxy, benzonitrile, o-hydroxy, o-hydoxybenzonitrile, 2-hydroxy-benzonitrileAfficher plus de résultatsAfficher moins de résultats
PubChem CID 11907
IUPAC Name 2-hydroxybenzonitrile
SMILES C1=CC=C(C(=C1)C#N)O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC161410050
FDS Afficher les documents
Thermo Scientific Chemicals
161410050
5 g Glass bottle N/A
Il en reste null
 
AC161410250
FDS Afficher les documents
Thermo Scientific Chemicals
161410250
25 g Glass bottle N/A
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications
Afficher plus de résultats
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-259-3

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only