CAS RN 845-10-3
Methyl Red Indicator, 0.1% (w/v) Alcoholic Solution, Ricca Chemical
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): Mixture InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Thermo Scientific Chemicals Methyl Red sodium salt, ACS
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: 2-[4-(Dimethylamino)phenyl]azobenzoic acid sodium salt PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Methyl Red Indicator, 1% (w/v), Ricca Chemical
For Acid-Base Titrations
Thermo Scientific Chemicals Methyl Red sodium salt
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: C.I. 13020; Methyl Red, water soluble PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Thermo Scientific Chemicals Methyl Red sodium salt, 0.02% w/v aq. soln.
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Methyl Red, Sodium Salt, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: C.I. 13020 PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Thermo Scientific Chemicals Methyl Red, sodium salt, pure, indicator
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt,C.I. 13020,MR PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Thermo Scientific Chemicals Methyl Red, sodium salt, ACS reagent
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt,C.I. 13020,MR PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Methyl Red Sodium Salt, ACS Reagent Grade, Ricca Chemical
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Methyl Red, 0.1% Aqueous, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Methyl Red Sodium Salt, ACS Grade, LabChem™
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Methyl Red, 1.0% Aqueous, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Methyl Red Sodium Salt, TCI America™
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: methyl red sodium salt,methyl red sodium,unii-311gz0qhwn,methyl purple solution,benzoic acid, 2-4-dimethylamino phenyl azo-, sodium salt,311gz0qhwn,4-dimethylaminoazobenzene-2'-carboxylic acid sodium salt,methyl red sodium salt c.i. 13020,eaetho masculinei 2,sodium methyl red PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
- 1
- 2