missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Red, sodium salt, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Clarity | (Aqueous) Passes Test |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Linear Formula | (CH3)2NC6H4N=NC6H4CO2Na |
| Visual Transition Interval | From pH 4.2 (pink) to pH 6.2 (yellow) |
| Beilstein | 16, 329 |
| Merck Index | 15, 6192 |
| Color Index No. | 13020 |
| Color | Orange/Red to Yellow |
| Quantity | 100 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Toxic if swallowed. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 212-682-9 |
| TSCA | TSCA |
RUO – Research Use Only