missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, sodium salt, pure, indicator

Catalog No. AC151415000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151415000 500 g Glass bottle
AC151410250 25 g Glass bottle
AC151411000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC151415000 Supplier Thermo Scientific Chemicals Supplier No. 151415000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00002426
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2NC6H4N=NC6H4CO2Na
Visual Transition Interval From pH 4.2 (red) to pH 6.2 (yellow)
Beilstein 16, 329
Merck Index 15, 6192
Color Index No. 13020
Color Beige or White to Yellow
Quantity 500 g
Physical Form Flakes
Formula Weight 291.27
Grade Pure
Chemical Name or Material Methyl Red, sodium salt, pure
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only