missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Sodium Salt, TCI America™

Catalog No. M04241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M04241G Supplier TCI America Supplier No. M04241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00002426
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym methyl red sodium salt, methyl red sodium, unii-311gz0qhwn, methyl purple solution, benzoic acid, 2-4-dimethylamino phenyl azo-, sodium salt, 311gz0qhwn, 4-dimethylaminoazobenzene-2'-carboxylic acid sodium salt, methyl red sodium salt c.i. 13020, eaetho masculinei 2, sodium methyl red
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Quantity 1 g
Formula Weight 291.29
Physical Form Crystalline Powder
Chemical Name or Material Methyl Red Sodium Salt