missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Indicator, 0.02% (w/v), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Methyl Red Indicator |
| CAS | 845-10-3, 7732-18-5 |
| Concentration | 0.02% (w/v) |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Color | Orange to Red |
| pH | 5 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Percent Purity | 0.02% W/V |
| Show More |