missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Indicator, 1% (w/v), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Methyl Red Indicator |
| CAS | 845-10-3, 7732-18-5 |
| Packaging | Natural Poly Bottle |
| pH Color Changes | pink (pH 4.2) - yellow (pH 6.2) |
| Solvent or Matrix | Aqueous solution |
| Color | Orange to Red |
| pH | 5 |
| Quantity | 120 mL |
| Percent Purity | 1% W/V |
| Grade | Indicator |
| Show More |