missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red, 0.1% Aqueous, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™
Specifications
Specifications
| CAS Max % | 99.9, 0.1 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Linear Formula | C15H14N3O2Na |
| Decomposition Information | Nitrogen oxides; Carbon monoxide; Carbon dioxide |
| Color | Orange |
| Solubility Information | Soluble in water |
| Quantity | 125 mL |
| Physical Form | Liquid |
| Formula Weight | 291.28 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |