missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Indicator, 0.1% (w/v) Alcoholic Solution, Ricca Chemical

Catalog No. SOD524
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SOD524 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SOD524 Supplier Ricca Chemical Company Supplier No. RSOD5200120C
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 845-10-3 , 65-56-1
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) Mixture
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Odor Characteristic alcohol
Packaging Amber Glass Bottle
Assay Percent Range 0.1% (w/v)
Color Red
Solubility Information Solubility in Water: Infinite
Quantity 120 mL
Physical Form Solution
Percent Purity 0.1% w/v
Grade Indicator
Chemical Name or Material Methyl Red
Recommended Storage Flammable