Learn More
Thermo Scientific Chemicals Methyl Red sodium salt, ACS
Description
Methyl Red sodium salt is used as a pH indicator. In microbiology, it is used to make agar plates in bacteriology. Further, it is used as a coloring agent and also finds use in mouthwash, toothpaste and in cosmetics. It is also employed as a rocket propellant. In addition to this, it is used in the preparation of reversible stain for rapid detection of stained cells in each specimen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Beilstein | 4065488 |
| Color | Brown to Red |
| Solubility Information | Soluble in water. Slightly soluble in ethanol. |
| Quantity | 25 g |
| Physical Form | Crystalline |
| Formula Weight | 291.29 |
| Grade | ACS |
| Chemical Name or Material | Methyl Red sodium salt |
Safety and Handling
| EINECSNumber | 212-682-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only