CAS RN 7772-98-7
Sodium Thiosulfate Anhydrous (Certified), Fisher Chemical™
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium sulfanidesulfonate SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate, 0.1N Standardized Solution
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate, 0.01N Standardized Solution
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate, 0.1 N standard solution
CAS: 7772-98-7 ∣ Na2O3S2 ∣ 158.10 g/mol
Sodium thiosulfate, anhydrous, 98+%
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium Thiosulfate, 0.20 Normal (N/5) Volumetric Solution, StandARd, Macron Fine Chemicals
CAS: 7772-98-7 Molecular Formula: H10Na2O8S2 Molecular Weight (g/mol): 248.17 MDL Number: MFCD00003499 InChI Key: PODWXQQNRWNDGD-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium pentahydrate sulfanidesulfonate SMILES: O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
Sodium Thiosulfate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium sulfanidesulfonate SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium Thiosulfate, DILUT-IT™ Analytical Concentrate, 0.1N, 1/10 equiv. = 15.81g Na2S2O3, J.T. Baker™
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium sulfanidesulfonate SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate concentrate, For 1L standard solution, 0.01 M Na2S2O3 (0.01N), Solstice
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate, 0.01N, Macron Fine Chemicals™
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 IUPAC Name: disodium sulfanidesulfonate SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium Thiosulfate, Anhydrous, Granular, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate, 99%, for analysis, anhydrous
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate, 1.0N Standardized Solution
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L PubChem CID: 24477 IUPAC Name: disodium sulfanidesulfonate SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
Sodium thiosulfate, 98.5%, extra pure, anhydrous
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L Synonym: sodium thiosulfate,sodium thiosulphate,disodium thiosulfate,sodium thiosulfate anhydrous,hypo,sodiumthiosulfate,chlorine cure,chlorine control,declor-it,thiosulfuric acid, disodium salt PubChem CID: 24477 SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
VeriSpec™ Thiosulphate - S2O32- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
CAS: 7772-98-7 Molecular Formula: Na2O3S2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00003499 InChI Key: AKHNMLFCWUSKQB-UHFFFAOYSA-L PubChem CID: 24477 IUPAC Name: disodium sulfanidesulfonate SMILES: [Na+].[Na+].[O-]S([S-])(=O)=O
- 1
- 2