missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 0.01N, Macron Fine Chemicals™

Catalog No. MKH3621
Change view
Click to view available options
Quantity:
1 L
Packaging:
narrow mouth polyethylene bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MKH3621 1 L narrow mouth polyethylene bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MKH3621 Supplier Avantor Supplier No. H36205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Color White
Physical Form Crystals
Quantity 1 L
Formula Weight 248.18
Packaging narrow mouth polyethylene bottle
Density 1.667g/cm3
Chemical Name or Material Sodium thiosulfate