missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.20 Normal (N/5) Volumetric Solution, StandARd, Macron Fine Chemicals
Specifications
Specifications
| Quantity | 1L |
| Specific Gravity | 1L = 1.06kg |
| Chemical Name or Material | Sodium Thiosulfate, 0.20 Normal (N/5) Volumetric Solution |