missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate concentrate, For 1L standard solution, 0.01 M Na2S2O3 (0.01N), Solstice

Catalog No. 60046709
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046709 6 x 1 Ea.
6002680 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046709 Supplier Solstice Supplier No. 382436X1EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for 1L standard solution, 0.01 M Na2S2O3 (0.01N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7772-98-7 , 7732-18-5
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Boiling Point 100°C
CAS Min % ≥90.0000%
Physical Form Concentrate
CAS Max % ≤100.0000%, ≤5.0000%
Quantity 6 x 1 Ea.
Packaging Ampoule
Density 1.0 g/cm3
Linear Formula Na2S2O3
UN Number NONH for all modes of transport
Formula Weight 158.11
Chemical Name or Material Sodium thiosulfate concentrate, For 1L standard solution, 0.01 M Na2S2O3 (0.01N)
Show More Show Less
ShelfLife 720 days from date of manufacture
Recommended Storage Room Temp