missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 99%, for analysis, anhydrous

Catalog No. AC450622500
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450622500 250 g
AC450620010 1 kg
AC450620050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450622500 Supplier Thermo Scientific Chemicals Supplier No. 450622500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7772-98-7 ∣ Na2O3S2 ∣ 158.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Melting Point 48.0°C
pH 6.0 to 8.4 (5% aq. soln.)
Physical Form Crystalline Powder
Quantity 250 g
Loss on Drying 1% max. (105°C)
Packaging Plastic Bottle
Assay Percent Range 98.5% min. (Iodimetry)
Linear Formula Na2S2O3
Merck Index 15,8821
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Formula Weight 158.1
Percent Purity 99%
Grade Analytical
Chemical Name or Material Sodium thiosulfate
Show More Show Less
EINECSNumber 231-867-5

RUO – Research Use Only