missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, DILUT-IT™ Analytical Concentrate, 0.1N, 1/10 equiv. = 15.81g Na2S2O3, J.T. Baker™

Catalog No. 02003955
Change view
Click to view available options
Quantity:
1 Pk.
Packaging:
Poly Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003955 1 Pk. Poly Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02003955 Supplier Avantor Supplier No. 469501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Quantity 1 Pk.
Packaging Poly Ampoule
Chemical Name or Material Sodium Thiosulfate, DILUT-IT™ Analytical Concentrate, 0.1N, 1/10 equiv. = 15.81g Na2S2O3