missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 1.0N Standardized Solution

Catalog No. AA35647K7
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35647K7 4 L
AA35647K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35647K7 Supplier Thermo Scientific Chemicals Supplier No. 035647K7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
PubChem CID 24477
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Physical Form Liquid
Color Colorless
Type Analytical Volumetric Solution
Quantity 4 L
Formula Weight 158.1
Grade Standard
Chemical Name or Material Sodium thiosulfate
EINECSNumber 231-867-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only